Sheep Neopterin ELISA Kit

Lieferant: ANTIBODIES.COM

A87013-96
ANTIA87013-96EA 700 EUR
ANTIA87013-96
Sheep Neopterin ELISA Kit
Tests ELISAs

Sheep Neopterin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of sheep Neopterin in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-to-use ELISA Kit
  • Detection range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%


Sheep Neopterin ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of sheep Neopterin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Neopterin antigen. During the incubation, Neopterin present in the samples or standards competes with the fixed amount of immobilised Neopterin for binding sites on the Biotinylated Anti-Neopterin Antibody. The more Neopterin present in a sample or standard, the less Biotinylated Anti-Neopterin Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Neopterin Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Neopterin present in each sample or standard. The concentration of Neopterin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Bestellinformation: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR