Anti-IDH1 Rabbit Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A92944-100
ANTIA92944-100EA 525 EUR
ANTIA92944-100
Anti-IDH1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to IDH1 for WB and IP with samples derived from Human, Mouse and Rat.
  • Validated applications: WB, IP
  • Recommended dilutions: WB: 1:500 to 1:2000, IP: 1:50 to 1:100
Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat