Actinomycin D 10 mmol/l in wässriger Lösung
Lieferant: G Bioscienes
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).
- Supplied as 50μl 10mM solution
G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.
Actinomycin D inhibits transcription by binding to DNA at the transcription initiation complex and therefore preventing elongation by the RNA polymerase.
Formel:
C₆₂H₈₆N₁₂O₁₆ |
MDL:
MFCD00005033 CAS-Nummer: 50-76-0 UN: 3462 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...