(S)-(+)-Camptothecin 2 mM
Lieferant: G Bioscienes
Synonyme:
(S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).
- Supplied as 1ml 2mM
G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.
Camptothecin is a cytotoxic quinoline alkaloid that inhibits the DNA enzyme topoisomerase I (Topo I).
Formel:
C₂₀H₁₆N₂O₄ |
MDL:
MFCD00081076 CAS-Nummer: 7689-03-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...