Dexamethason 100 mM
Lieferant: G Bioscienes
Synonyme:
9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).
- Supplied as 1ml 10mM solution.
G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.
Dexamethasone is a potent and stable glucocorticoid steroid hormone that is often used as an anti-inflammatory and anti-rheumatic drug.
The mechanism of action is not fully understood, however it is thought that it may inhibit prostaglandin synthesis.
Formel:
C₂₂H₂₉FO₅ Molecular Weight: 392,47 g/mol |
MDL:
MFCD00064136 CAS-Nummer: 50-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...