Doxorubicin 2 mM
Lieferant: G Bioscienes
Synonyme:
Adriamycin, (7S,9S)-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione, Doxil, Hydroxydaunorubicin, Adriablastin, Doxorubicinum, Doxorubicin, Myocet
Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).
- Supplied as 50μl 2mM solution.
G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.
Doxorubicin (or hydroxyldaunorubicin) is an anthracycline antibiotic that intercalates DNA, inhibiting the unwinding of DNA by topoisomerase II.
Formel:
C₂₇H₂₉NO₁₁ Molecular Weight: 543,52 g/mol |
CAS-Nummer:
23214-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...