Blades
Lieferant: APOLLO HERCKENRATH
10-210-090
APOL10-210-090EA 463 EUR
APOL10-210-090
Blades
Rasiermesser und Klingen
These blades made from carbon steel feature a handle bar made from steel.
Verpackung: 5 blades per bag.