D(-)-Sorbit

Lieferant: Apollo Scientific
Achtung

Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol

BI2889-1KG BI2889-500G
APOSBI2889-1KGEA 0 EUR
APOSBI2889-1KG APOSBI2889-500G
D(-)-Sorbit
D(-)-Sorbit
Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Lagertemperatur: Raumtemperatur
MDL: MFCD00004708
CAS-Nummer: 50-70-4
Merck Index: 13,08797

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR