2-(Cyclohexylamino)ethansulfonsäure (CHES)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Cyclohexyltaurine, CHES
BI3458-100G
APOSBI3458-100GEA
26.1
EUR
APOSBI3458-100G
2-(Cyclohexylamino)ethansulfonsäure (CHES)
2-(Cyclohexylamino)ethansulfonsäure (CHES)
- Assay (by titration) 98+%
- Water (by Karl Fischer) <1.0%
- Biological buffer - useful range 8.6-10.0
Formel:
C₈H₁₇NO₃S Molecular Weight: 207,29 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003835 CAS-Nummer: 103-47-9 EINECS: 203-115-6 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...