Teicoplanin

Lieferant: Apollo Scientific
Achtung

BI3859-100MG
APOSBI3859-100MGEA 168 EUR
APOSBI3859-100MG
Teicoplanin
Teicoplanin
Formel: C₈₉H₉₉Cl₂N₉O₃₃
Lagertemperatur: Gefrierschrank
MDL: MFCD05664625
CAS-Nummer: 61036-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR