Hemin chloride ≥96%

Lieferant: Apollo Scientific

Synonyme: Chlorohemin, 1,3,5,8-Tetramethyl-2,4-divinylporphine-6,7-dipropionic acid ferrichloride, Chloroprotoporphyrin IX iron(III), Haemin, Hemin, Ferriprotoporphyrin IX chloride

BI4570-10G
APOSBI4570-10GEA 818 EUR
APOSBI4570-10G
Hemin chloride ≥96%
Hemin chloride

Bitte beachten Use only outdoors or in a well-ventilated area. Avoid breathing dust/fumes. Avoid release to the environment. Wear protective gloves. Wash all exposed external body areas thoroughly after handling. If in eyes rinse cautiously with hazard eye protection and face protection.

Formel: C₃₄H₃₂ClFeN₄O₄
Molecular Weight: 651,95 g/mol
Schmelzpunkt: 46,..53 °C
MDL: MFCD00010726
CAS-Nummer: 16009-13-5

Bestellen

Spezifikation Testergebnisse

Purity 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR