2,2'-Bicinchoninic acid disodium salt ≥98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Sodium [2,2'-biquinoline]-4,4'-dicarboxylate, BCA disodium salt, Disodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-Biquinoline-4,4'-dicarboxylic acid disodium salt, Disodium 2,2'-Biquinoline-4,4'-dicarboxylate, Bicinchoninic acid disodium salt

BI5837-25G BI5837-5G BI5837-1G
APOSBI5837-25GEA 248 EUR
APOSBI5837-25G APOSBI5837-5G APOSBI5837-1G
2,2'-Bicinchoninic acid disodium salt ≥98%
2,2'-Bicinchoninic acid disodium salt

Used in the determination of protein concentration. Proteins reduce alkaline Cu(II) to Cu(I) which complexes with BCA to form a purple complex. Absorbance at 562nm is directly proportional to protein concentration.

Formel: C₂₀H₁₀N₂Na₂O₄
Molecular Weight: 388.29 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037500
CAS-Nummer: 979-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR