3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS

BI5849-5G
APOSBI5849-5GEA 21.9 EUR
APOSBI5849-5G
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz ≥98%
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz

Enzyme test reagent.


  • Assay (by titration) 98+%
  • Water (by Karl Fischer) < 3.0%

Formel: C₆H₃Cl₂NaO₄S
Molecular Weight: 265,05 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009798
CAS-Nummer: 54970-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR