1,3-Phenylendimethanthiol 98%

Lieferant: Apollo Scientific

Synonyme: 1,3-Bis(mercaptomethyl)benzene, alpha,alpha'-Dimercapto-m-xylene

BI7567-5G
APOSBI7567-5GEA 132 EUR
APOSBI7567-5G
1,3-Phenylendimethanthiol 98%
1,3-Benzenedimethanethiol
Formel: C₈H₁₀S₂
Molecular Weight: 170.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012311
CAS-Nummer: 41563-69-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR