Chlortetracyclin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
BI7654-10G
BI7654-100G
BI7654-25G
APOSBI7654-10GEA
50.9
EUR
APOSBI7654-10G
APOSBI7654-100G
APOSBI7654-25G
Chlortetracyclin Hydrochlorid
Chlortetracyclin Hydrochlorid
Formel:
C₂₂H₂₃ClN₂O₈·HCl Schmelzpunkt: 210 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD26142699 CAS-Nummer: 64-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...