Tricin ≥99%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Tris(hydroxymethyl)methyl)glycin

BI9678T-100G BI9678T-1KG BI9678T-500G
APOSBI9678T-100GEA 36.4 EUR
APOSBI9678T-100G APOSBI9678T-1KG APOSBI9678T-500G
Tricin ≥99%
Tricin

Component for separation of low molecular weight peptides. Biological buffer - useful range 7.4-8.8

Formel: (HOCH₂)₃CNHCH₂CO₂H
Schmelzpunkt: 187 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004277
CAS-Nummer: 5704-04-1
EINECS: 227-193-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR