Ampicillin Natriumsalz

Lieferant: Apollo Scientific
Gefahr

BIA0104-100G BIA0104-25G BIA0104-250G
APOSBIA0104-100GEA 0 EUR
APOSBIA0104-100G APOSBIA0104-25G APOSBIA0104-250G
Ampicillin Natriumsalz
Ampicillin Natriumsalz

Inhibitor of bacterial cell wall synthesis.


  • Cell culture tested

Formel: C₁₆H₁₈N₃NaO₄S
Lagertemperatur: Kühlschrank
MDL: MFCD00064313
CAS-Nummer: 69-52-3
EINECS: 200-708-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR