(R)-tert-Butyl-2-aminopropanoate hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
D-Alanine tert-butyl ester hydrochloride, tert-Butyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride, H-D-Ala-OtBu.HCl, H-D-Ala-OtBu·HCl
BIA0105-1G
APOSBIA0105-1GEA
21.9
EUR
APOSBIA0105-1G
(R)-tert-Butyl-2-aminopropanoate hydrochloride
(R)-tert-Butyl-2-aminopropanoate hydrochloride
USP, pyrogen free.
Formel:
C₇H₁₅NO₂·HCl Molecular Weight: 181,7 g/mol Schmelzpunkt: 170…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077087 CAS-Nummer: 59531-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...