L-Alanin-ethylester-hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Ala-OEt.HCl, Ethyl (S)-2-Aminopropionate Hydrochloride, L-Ethyl alanate hydrochloride, (S)-2-Aminopropionic Acid Ethyl Ester Hydrochloride, H-Ala-OEt·HCl
BIA0106-25G
APOSBIA0106-25GEA
21.9
EUR
APOSBIA0106-25G
L-Alanin-ethylester-hydrochlorid
L-Alanine ethyl ester hydrochloride
Formel:
C₅H₁₁NO₂ Molecular Weight: 153.61 g/mol Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063662 CAS-Nummer: 1115-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...