Apramycin sulphate

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2R,3R,4S,5S,6S)-2-[[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy-4-hydroxy-3-(methylamino)-2,3,4,4a,6,7,8,8a-octahydropyrano[3,2-b]pyran-2-yl]oxy]-5-amino-6-(hydroxymethyl)oxane-3,4-diol, sulphuric acid, Nebramycin II, Apramycin sulphate salt, (2R,3R,4S,5S,6S)-5-Amino-2-(((2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-(((1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl)oxy)-4-hydroxy-3-(methylamino)octahydropyrano[3,2-b]pyran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol xsulphate, Nebramycin II sulphate

BIA0164-25G BIA0164-5G
APOSBIA0164-25GEA 270 EUR
APOSBIA0164-25G APOSBIA0164-5G
Apramycin sulphate
Apramycin sulphate

An aminoglycoside antibiotic with bactericidal activity

Formel: C₂₁H₄₁N₅O₁₁·H₂SO₄
Molecular Weight: 637.65 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06200257
CAS-Nummer: 65710-07-8
EINECS: 265-890-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR