Actinomycin D ≥99%

Lieferant: Apollo Scientific
Gefahr

BIA0175-10MG BIA0175-25MG BIA0175-50MG
APOSBIA0175-10MGEA 183 EUR
APOSBIA0175-10MG APOSBIA0175-25MG APOSBIA0175-50MG
Actinomycin D ≥99%
Actinomycin D

Antineoplastic antibiotic


  • Purity: ≥99%

Formel: C₆₂H₈₆N₁₂O₁₆
Schmelzpunkt: 251…253 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005033
CAS-Nummer: 50-76-0
EINECS: 200-063-6
UN: 3462
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR