ADA ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Carbamoylmethyliminodi(essigsäure), N-(2-Acetamido)iminodiessigsäure
BIA0302-500G
APOSBIA0302-500GEA
62.5
EUR
APOSBIA0302-500G
ADA ≥98%
ADA
Biological buffer - useful range 6.0 - 7.2
Formel:
H₂NC(O)CH₂N(CH₂COOH)₂ Schmelzpunkt: 219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008031 CAS-Nummer: 26239-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...