Adeninsulfat ≥99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Adeninhemisulfat

BIA0543-100G BIA0543-25G BIA0543-500G
APOSBIA0543-100GEA 109 EUR
APOSBIA0543-100G APOSBIA0543-25G APOSBIA0543-500G
Adeninsulfat ≥99%
Adeninsulfat

Cytokin growth regulatorPlant tissue culture tested.

Formel: C₅H₅N₅·½H₂SO₄
Schmelzpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR