D(+)-Prolin

Lieferant: Apollo Scientific
Achtung

BIA0718-100G BIA0718-25G
APOSBIA0718-100GEA 0 EUR
APOSBIA0718-100G APOSBIA0718-25G
D(+)-Prolin
D(+)-Prolin
Formel: C₅H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00064317
CAS-Nummer: 344-25-2
EINECS: 206-452-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR