D-Alanin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-(-)-2-Aminopropionic acid, Alanin, D-α-Alanin, D(-)-Alanin, (R)-(-)-Alanin, H-D-Ala-OH, α-Alanin, (R)-(-)-2-Aminopropansäure, D(-)-2-Aminopropionic acid, (R)-(-)-2-Aminopropionsäure
BIA0863-25G
APOSBIA0863-25GEA
21.9
EUR
APOSBIA0863-25G
D-Alanin
D-Alanin
Chiral intermediate for pharmaceuticals and agrochemicals.
- Materials for peptides
- USP
- Pyrogen free
Formel:
C₃H₇NO₂ Molecular Weight: 89.09 g/mol Siedepunkt: 189 Schmelzpunkt: 291…295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008077 CAS-Nummer: 338-69-2 EINECS: 206-418-1 Merck Index: 13,00203 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...