D(+)-Saccharose Pure BP, Ph. Eur., JP, USP, pharma grade
Lieferant: Apollo Scientific
Synonyme:
Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker
BIA1125-1KG
BIA1125-5KG
APOSBIA1125-1KGEA
65.4
EUR
APOSBIA1125-1KG
APOSBIA1125-5KG
D(+)-Saccharose Pure BP, Ph. Eur., JP, USP, pharma grade
D(+)-Saccharose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342.3 g/mol Schmelzpunkt: 186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006626 CAS-Nummer: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...