Acridinorange (Zinkchlorid-Doppelsalz)

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

BIA1145-100G BIA1145-25G
APOSBIA1145-100GEA 79.9 EUR
APOSBIA1145-100G APOSBIA1145-25G
Acridinorange (Zinkchlorid-Doppelsalz)
Acridinorange (Zinkchlorid-Doppelsalz)

A fluorescent stain for nuclei acids in agarose andpolyacrylamide gels.

Formel: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Molecular Weight: 369.96 g/mol
Schmelzpunkt: 165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081043
CAS-Nummer: 10127-02-3
UN: 3143
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR