Dabsylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
DABS-Cl, 4-Dimethylaminoazobenzene-4'-sulphonyl Chloride, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonyl chloride
BIA115-500MG
APOSBIA115-500MGEA
87
EUR
APOSBIA115-500MG
Dabsylchlorid
Dabsyl chloride
Used as a chromophoric label and for HPLC separation.
Formel:
C₁₄H₁₄ClN₃O₂S Molecular Weight: 323.80 g/mol Schmelzpunkt: 185 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007444 CAS-Nummer: 56512-49-3 EINECS: 260-235-1 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...