Agarose Molekularsieb-Qualität

Lieferant: Apollo Scientific
Achtung

BIA1197-100G BIA1197-250G BIA1197-50G
APOSBIA1197-100GEA 0 EUR
APOSBIA1197-100G APOSBIA1197-250G APOSBIA1197-50G
Agarose Molekularsieb-Qualität
Agarose

Agarose Molecular Screening 8 for improved resolution of DNA fragments 50-1000bp, especially primers. Higher gel strength than agarose MS6, suitable for use in blotting.


  • Gel strength at 3% >1500g/cm2 Gelling temp. 3% <35.5°C Melting temp. 3% <80°C Moisture <7% Ash <0.35% EEO <0.12 Sulphate <0.11%. DNAse/RNAse activity not detected.

Lagertemperatur: Raumtemperatur
MDL: MFCD00081294
CAS-Nummer: 9012-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR