N-Acetyl-L-alanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ac-Ala-OH, Acetyl-L-alanine
BIA1219-25G
APOSBIA1219-25GEA
21.9
EUR
APOSBIA1219-25G
N-Acetyl-L-alanine
N-Acetyl-L-alanine
Formel:
C₅H₉NO₃ Molecular Weight: 131,131 g/mol Schmelzpunkt: 125…126 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00063132 CAS-Nummer: 97-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...