2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose
Lieferant: Apollo Scientific
BIA1238-5G
APOSBIA1238-5GEA
0
EUR
APOSBIA1238-5G
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-glucopyranose
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-beta-D-glucopyranose
Formel:
C₁₆H₂₃NO₁₀ Schmelzpunkt: 186…189 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006595 CAS-Nummer: 7772-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...