1,4-Phenylendiisothiocyanat

Lieferant: Apollo Scientific
Gefahr

Synonyme: p-Phenylendiisothiocyanat, Bitoscanat, p-Diisothiocyanatobenzol, 1,4-Phenyldiisothiocyanat, 1,4-Diisothiocyanatobenzol

BIA130-1G
APOSBIA130-1GEA 27.7 EUR
APOSBIA130-1G
1,4-Phenylendiisothiocyanat
1,4-Phenylendiisothiocyanat

Solid phase protein sequencingRef: Laursen,R.A et al (1972) FEBS lett 21 67

Formel: C₈H₄N₂S₂
Molecular Weight: 192,27 g/mol
Schmelzpunkt: 128…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004811
CAS-Nummer: 4044-65-9
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR