Nɑ-Acetyl-L-glutamine

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Acetyl-L-glutamine, N-Ac-Gln-OH, Ac-L-Gln-OH, Ac-Gln-OH

BIA1740-100G
APOSBIA1740-100GEA 37.7 EUR
APOSBIA1740-100G
Nɑ-Acetyl-L-glutamine
Nalpha-Acetyl-L-glutamine
Formel: C₇H₁₂N₂O₄
Molecular Weight: 188,18 g/mol
Schmelzpunkt: 206…208 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00038159
CAS-Nummer: 2490-97-3
EINECS: 219-647-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR