N-Acetyl-DL-Tryptophan

Lieferant: Apollo Scientific

BIA1791-100G
APOSBIA1791-100GEA 226 EUR
APOSBIA1791-100G
N-Acetyl-DL-Tryptophan
N-Acetyl-DL-Tryptophan
Formel: C₁₃H₁₄N₂O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00005644
CAS-Nummer: 87-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR