15-Acetoxyscirpenol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
15-Acetoxyscirpendiol, 15-Acetoxyscirpen-3,4-diol
BIA2304-1MG
APOSBIA2304-1MGEA
133
EUR
APOSBIA2304-1MG
15-Acetoxyscirpenol
15-Acetoxyscirpenol
Formel:
C₁₈H₂₆O₅ Molecular Weight: 322.396 g/mol Schmelzpunkt: 165…167 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00078774 CAS-Nummer: 2623-22-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...