7-Amino-4-methylcumarin
Lieferant: Apollo Scientific
|
Achtung
|
BIA3001-1G
BIA3001-500MG
BIA3001-5G
APOSBIA3001-1GEA
0
EUR
APOSBIA3001-1G
APOSBIA3001-500MG
APOSBIA3001-5G
7-Amino-4-methylcumarin
7-Amino-4-methylcumarin
Formel:
C₁₀H₉NO₂ Schmelzpunkt: 223…226 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006868 CAS-Nummer: 26093-31-2 EINECS: 247-454-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...