Adipic acid monobenzyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Benzyloxy)-6-oxohexanoic acid

BIA4002-1G
APOSBIA4002-1GEA 21.9 EUR
APOSBIA4002-1G
Adipic acid monobenzyl ester
Adipic acid monobenzyl ester
Formel: C₁₃H₁₆O₄
Molecular Weight: 236,264 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00441728
CAS-Nummer: 40542-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR