4-Aminophenylphosphate sodium salt

Lieferant: Apollo Scientific
Achtung

BIA4012-100MG BIA4012-1G BIA4012-500MG
APOSBIA4012-100MGEA 0 EUR
APOSBIA4012-100MG APOSBIA4012-1G APOSBIA4012-500MG
4-Aminophenylphosphate sodium salt
4-Aminophenylphosphate sodium salt
Formel: C₆H₇NO₄P·Na·4.0H₂O
Schmelzpunkt: 176 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 108084-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR