7-Aminoactinomycin (7-AAD)

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,7-Diamino-N,N'-bis[(6S,9R,10S,13R,18aS)-6,13-diisopropyl-2,5,9-trimethyl-1,4,7,11,14-pentaoxohexadecahydro-1H-pyrrolo[2,1-i][1,4,7,10,13]oxatetraazacyclohexadecin-10-yl]-4,6-dimethyl-3-oxo-3H-phenox azine-1,9-dicarboxamide, Actinomy, 7-Aminoactinomycin D, NSC-239759, NSC 239759, 2,7-diamino-4,6-dimethyl-3-oxo-1-N,9-N-bis[7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]phenoxazine-1,9-dicarboxamide, 7-AAD

BIA4306-1MG BIA4306-5MG
APOSBIA4306-1MGEA 183 EUR
APOSBIA4306-1MG APOSBIA4306-5MG
7-Aminoactinomycin (7-AAD)
7-Aminoactinomycin D (7-AAD)

Fluorescent DNA stain.

Formel: C₆₂H₈₇N₁₃O₁₆
Molecular Weight: 1270.43 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077295
CAS-Nummer: 7240-37-1
UN: 3462
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR