Anisomycin (aus Streptomyces griseolus)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
BIA4315-250MG
APOSBIA4315-250MGEA
132
EUR
APOSBIA4315-250MG
Anisomycin (aus Streptomyces griseolus)
Anisomycin
Formel:
C₁₄H₁₉NO₄ Molecular Weight: 265,31 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077650 CAS-Nummer: 22862-76-6 EINECS: 245-269-7 UN: 3462 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...