Apicidin

Lieferant: Apollo Scientific
Gefahr

Synonyme: cyclo-[L-(2-Amino-8-oxodecanoyl)-L-(N-methoxy-tryptophan)-L-isoleucyl-D-pipecolinyl]

BIA4318-1MG
APOSBIA4318-1MGEA 27.7 EUR
APOSBIA4318-1MG
Apicidin
Apicidin
Formel: C₃₄H₄₉N₅O₆
Molecular Weight: 623,8 g/mol
Schmelzpunkt: 190 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 183506-66-3
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR