Acetobromo-α-D-glucuronic acid methyl ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: BROMOMETHYLTRIACETYL GLUCURONATE, Acetobromo-α-D-glucuronic Acid Methyl Ester, Methyl acetobromo-α-D-glucuronate, (2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

BIA6200-1G BIA6200-5G BIA6200-25G
APOSBIA6200-1GEA 50.9 EUR
APOSBIA6200-1G APOSBIA6200-5G APOSBIA6200-25G
Acetobromo-α-D-glucuronic acid methyl ester 98%
Acetobromo-alpha-D-glucuronic acid methyl ester

Stabilised with 2% calcium carbonate

Formel: C₁₃H₁₇BrO₉
Molecular Weight: 397.174 g/mol
Schmelzpunkt: ∼ 105 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00061613
CAS-Nummer: 21085-72-3
EINECS: 244-203-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR