Cetrimoniumbromid

Lieferant: Apollo Scientific

Synonyme: Hexadecyltrimethylammoniumbromid, N-Cetyl-N,N,N-trimethylammoniumbromid, CTAB, N-Hexadecyl-N,N,N-trimethylammoniumbromid, Cetyltrimethylammoniumbromid

BIA6284-500G
APOSBIA6284-500GEA 33.5 EUR
APOSBIA6284-500G
Cetrimoniumbromid
Cetrimoniumbromid
Formel: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Molecular Weight: 364,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011772
CAS-Nummer: 57-09-0
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR