Bilirubin

Lieferant: Apollo Scientific

Synonyme: 3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

BIB0134-1G
APOSBIB0134-1GEA 97.3 EUR
APOSBIB0134-1G
Bilirubin
Bilirubin
Formel: C₃₃H₃₆N₄O₆
Molecular Weight: 584,67 g/mol
Schmelzpunkt: 192 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005499
CAS-Nummer: 635-65-4
EINECS: 211-239-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR