D(+)-Biotin 99%

Lieferant: Apollo Scientific
Achtung

BIB0603-1G BIB0603-5G BIB0603-10G
APOSBIB0603-1GEA 0 EUR
APOSBIB0603-1G APOSBIB0603-5G APOSBIB0603-10G
D(+)-Biotin 99%
D(+)-Biotin
Formel: C₁₀H₁₆N₂O₃S
Schmelzpunkt: 231…233 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005541
CAS-Nummer: 58-85-5
EINECS: 200-399-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR