Hydroxynaphtholblau
Lieferant: Apollo Scientific
Synonyme:
1-(2-Hydroxy-4-sulpho-1-naphthylazo)-2-naphthol-3,6-disulphonic acid trisodium salt
BIB1006-10G
APOSBIB1006-10GEA
94.4
EUR
APOSBIB1006-10G
Hydroxynaphtholblau
Hydroxynaphthol blue
Formel:
C₂₀H₁₁N₂Na₃O₁₁S₃ Molecular Weight: 620,47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036393 CAS-Nummer: 63451-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...