Calcein
Lieferant: Apollo Scientific
BIB1007-1G
BIB1007-5G
APOSBIB1007-1GEA
58
EUR
APOSBIB1007-1G
APOSBIB1007-5G
Calcein
Calcein
Formel:
C₃₀H₂₆N₂O₁₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005049 CAS-Nummer: 154071-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...