Sulpho-NHS-LC-Biotin

Lieferant: Apollo Scientific
Gefahr

Synonyme: Sulphosuccinimidyl-6-(biotinamido)hexanoate, Biotinamidohexanoic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt

BIB103-100MG
APOSBIB103-100MGEA 306 EUR
APOSBIB103-100MG
Sulpho-NHS-LC-Biotin
Sulpho-NHS-LC-Biotin

Water soluble, used in sandwich ELISAs and in In Vivo tumor site localisationRef: Suter.M & Butler J.E (1986) Immunology letters 13, 313-316

Formel: C₂₀H₂₉N₄NaO₉S₂
Molecular Weight: 556,59 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00078534
CAS-Nummer: 127062-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR