Sulphosuccinimidyl-2-(biotinamido)ethyl-1,3-dithiopropionate

Lieferant: Apollo Scientific
Achtung

Synonyme: Sulpho NHS SS biotin

BIB105-100MG
APOSBIB105-100MGEA 102 EUR
APOSBIB105-100MG
Sulphosuccinimidyl-2-(biotinamido)ethyl-1,3-dithiopropionate
Sulphosuccinimidyl-2-(biotinamido)ethyl-1,3-dithiopropionate

Reversible biotinylation -Water solubleProtein Isolation -Recover Protein-DNA


Ref: Mouton, C.A. et.al. (1982) Arch. Biochem. Biophys. 218, 101-108

Formel: C₂₂H₃₂N₄O₁₁S₄
Molecular Weight: 656,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861945
CAS-Nummer: 858128-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR