N-Iodoacetyl-N-biotinylhexylenediamine

Lieferant: Apollo Scientific
Achtung

Synonyme: Iodoacetyl LC biotin

BIB107-100MG
APOSBIB107-100MGEA 275 EUR
APOSBIB107-100MG
N-Iodoacetyl-N-biotinylhexylenediamine
N-Iodoacetyl-N-biotinylhexylenediamine

Sulphhydryl reactive -Spacer arm


Ref: Suton,K.et al.(1984) J.Mol.Bio.178, 323-339

Formel: C₁₈H₃₁IN₄O₃S
Molecular Weight: 510,433 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00054987
CAS-Nummer: 93285-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR